Product Details
- SNP ID
-
rs7628368
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:113005772 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTTCTATGCACAGTACAGTGTG[A/G]GAGTCTCCACTTAGCACTGGTATCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GTPBP8
PubMed Links
Gene Details
- Gene
- GTPBP8
- Gene Name
- GTP binding protein 8 (putative)
There are no transcripts associated with this gene.
- Gene
- NEPRO
- Gene Name
- nucleolus and neural progenitor protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319109.1 |
1255 |
Missense Mutation |
CCA,TCA |
P420S |
NP_001306038.1 |
NM_001319110.1 |
1255 |
Missense Mutation |
CCA,TCA |
P323S |
NP_001306039.1 |
NM_001319111.1 |
1255 |
Missense Mutation |
CCA,TCA |
P287S |
NP_001306040.1 |
NM_001319112.1 |
1255 |
Missense Mutation |
CCA,TCA |
P379S |
NP_001306041.1 |
NM_001319114.1 |
1255 |
Missense Mutation |
CCA,TCA |
P354S |
NP_001306043.1 |
NM_001319115.1 |
1255 |
Missense Mutation |
CCA,TCA |
P393S |
NP_001306044.1 |
NM_015412.3 |
1255 |
Missense Mutation |
CCA,TCA |
P490S |
NP_056227.2 |
XM_005247276.2 |
1255 |
Missense Mutation |
CCA,TCA |
P354S |
XP_005247333.1 |
XM_011512635.2 |
1255 |
Missense Mutation |
CCA,TCA |
P287S |
XP_011510937.1 |
XM_011512636.2 |
1255 |
Missense Mutation |
CCA,TCA |
P287S |
XP_011510938.1 |
XM_011512638.2 |
1255 |
Missense Mutation |
CCA,TCA |
P287S |
XP_011510940.1 |
XM_017006102.1 |
1255 |
UTR 3 |
|
|
XP_016861591.1 |
XM_017006103.1 |
1255 |
UTR 3 |
|
|
XP_016861592.1 |
View Full Product Details