Product Details

SNP ID
rs78778792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44144560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTTTGTGTCTTCCATAGTGT[G/T]CCAGGCACGATTCTGCCTTCTCTCA
Phenotype
MIM: 604750
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF225 PubMed Links

Gene Details

Gene
ZNF225
Gene Name
zinc finger protein 225
There are no transcripts associated with this gene.

Gene
ZNF234
Gene Name
zinc finger protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144824.1 346 UTR 5 NP_001138296.1
NM_006630.2 346 UTR 5 NP_006621.1
XM_006722974.3 346 UTR 5 XP_006723037.1
XM_017026149.1 346 UTR 5 XP_016881638.1
XM_017026150.1 346 UTR 5 XP_016881639.1
XM_017026151.1 346 UTR 5 XP_016881640.1
XM_017026152.1 346 UTR 5 XP_016881641.1
XM_017026153.1 346 UTR 5 XP_016881642.1

View Full Product Details