Product Details

SNP ID
rs2286415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74034331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAGAAGACAGACTGGATGCCAA[A/C]GGTCTTGACTTTAGACTTCTCTTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BBOF1 PubMed Links

Gene Details

Gene
BBOF1
Gene Name
basal body orientation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025057.2 Intron NP_079333.2
XM_005268092.3 Intron XP_005268149.1
XM_005268097.3 Intron XP_005268154.1
XM_005268098.4 Intron XP_005268155.1
XM_011537170.2 Intron XP_011535472.1
XM_011537171.2 Intron XP_011535473.1
XM_011537172.1 Intron XP_011535474.1
XM_011537174.2 Intron XP_011535476.1
XM_011537175.2 Intron XP_011535477.1
XM_011537176.2 Intron XP_011535478.1
XM_011537177.2 Intron XP_011535479.1
XM_011537178.2 Intron XP_011535480.1
XM_011537179.2 Intron XP_011535481.1
XM_017021659.1 Intron XP_016877148.1
XM_017021660.1 Intron XP_016877149.1
XM_017021661.1 Intron XP_016877150.1
XM_017021662.1 Intron XP_016877151.1
XM_017021663.1 Intron XP_016877152.1

View Full Product Details