Product Details

SNP ID
rs34373915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12675556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTGGCCCTGCCTGTGGGTTCC[A/G]GTGTGGTGCAGTCGCTGGCCTACCA
Phenotype
MIM: 600944 MIM: 609458 MIM: 616850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHPS PubMed Links

Gene Details

Gene
DHPS
Gene Name
deoxyhypusine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206974.1 1340 Intron NP_001193903.1
NM_001930.3 1340 Intron NP_001921.1
NM_013406.2 1340 Intron NP_037538.1
XM_011527770.1 1340 UTR 3 XP_011526072.1
XM_011527771.2 1340 Intron XP_011526073.1
Gene
MAN2B1
Gene Name
mannosidase alpha class 2B member 1
There are no transcripts associated with this gene.

Gene
WDR83
Gene Name
WD repeat domain 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099737.2 1340 Missense Mutation AGT,GGT S278G NP_001093207.1
NM_032332.3 1340 Missense Mutation AGT,GGT S278G NP_115708.1
Gene
WDR83OS
Gene Name
WD repeat domain 83 opposite strand
There are no transcripts associated with this gene.

View Full Product Details