Product Details

SNP ID
rs17505250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37090105 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGCTTAGCTGAAGGCACTAACA[A/G]CCTAGATGGATGTGGCTTACAACAG
Phenotype
MIM: 176871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2AK2 PubMed Links

Gene Details

Gene
EIF2AK2
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
There are no transcripts associated with this gene.

Gene
GPATCH11
Gene Name
G-patch domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278505.1 Intron NP_001265434.1
NM_001322249.1 Intron NP_001309178.1
NM_174931.3 Intron NP_777591.3
XM_017003748.1 Intron XP_016859237.1
Gene
HEATR5B
Gene Name
HEAT repeat containing 5B
There are no transcripts associated with this gene.

View Full Product Details