Product Details

SNP ID
rs61740026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33092900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTACCCAGCCTCATGGGTATCCC[A/G]TAGTAGGCTGTGCCTGGGCTGACAT
Phenotype
MIM: 611458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLB1 PubMed Links

Gene Details

Gene
GLB1
Gene Name
galactosidase beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000404.3 1487 Intron NP_000395.2
NM_001079811.2 1487 Intron NP_001073279.1
NM_001135602.2 1487 Intron NP_001129074.1
NM_001317040.1 1487 Intron NP_001303969.1
Gene
TMPPE
Gene Name
transmembrane protein with metallophosphoesterase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039770.2 1487 Silent Mutation TAC,TAT Y432Y NP_001034859.2
NM_001136238.1 1487 Silent Mutation TAC,TAT Y295Y NP_001129710.1

View Full Product Details