Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001303270.1 | 1272 | Intron | NP_001290199.1 | ||
NM_020877.3 | 1272 | UTR 5 | NP_065928.2 | ||
XM_011523663.1 | 1272 | Intron | XP_011521965.1 | ||
XM_011523664.2 | 1272 | Intron | XP_011521966.1 | ||
XM_011523666.1 | 1272 | Intron | XP_011521968.1 | ||
XM_011523667.2 | 1272 | Intron | XP_011521969.1 | ||
XM_011523668.1 | 1272 | Intron | XP_011521970.1 | ||
XM_011523669.2 | 1272 | Intron | XP_011521971.1 | ||
XM_011523670.2 | 1272 | Intron | XP_011521972.1 | ||
XM_017024218.1 | 1272 | Intron | XP_016879707.1 | ||
XM_017024219.1 | 1272 | Intron | XP_016879708.1 |