Product Details

SNP ID
rs41413045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46203247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTGGCCTCCTATGGTCTGAGT[C/T]AGAACCCAGCAGAGAGTTCATGCTC
Phenotype
MIM: 601159 MIM: 601268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR1 PubMed Links

Gene Details

Gene
CCR1
Gene Name
C-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001295.2 1138 Silent Mutation TAA,TGA *356* NP_001286.1
Gene
CCR3
Gene Name
C-C motif chemokine receptor 3
There are no transcripts associated with this gene.

View Full Product Details