Product Details
- SNP ID
-
rs114635197
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51172494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCATGTGGCCACACTTACAGGTC[C/T]TTGTCTCTTCACTCAATGTCAGTTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928517
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139750636] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928517
- Gene Name
- uncharacterized LOC101928517
There are no transcripts associated with this gene.
- Gene
- SIGLEC17P
- Gene Name
- sialic acid binding Ig like lectin 17, pseudogene
There are no transcripts associated with this gene.
View Full Product Details