Product Details

SNP ID
rs34893204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44775266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTCTTTAGTAATGAAGGTGA[A/T]GCCATCAAAGTTTTTGTGCGAATTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF15 PubMed Links

Gene Details

Gene
KIF15
Gene Name
kinesin family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020242.2 227 Missense Mutation GAA,GAT E25D NP_064627.1
XM_006713264.3 227 Intron XP_006713327.1
XM_011533964.2 227 Missense Mutation GAA,GAT E25D XP_011532266.1
XM_017006884.1 227 Missense Mutation GAA,GAT E25D XP_016862373.1
XM_017006885.1 227 Missense Mutation AAG,ATG K12M XP_016862374.1
XM_017006886.1 227 UTR 5 XP_016862375.1
XM_017006887.1 227 Missense Mutation GAA,GAT E25D XP_016862376.1
XM_017006888.1 227 Intron XP_016862377.1
XM_017006889.1 227 Missense Mutation GAA,GAT E25D XP_016862378.1
XM_017006890.1 227 Intron XP_016862379.1

View Full Product Details