Product Details

SNP ID
rs4638862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45840651 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCTCCCTGCAGCTCTACACCCTC[A/G]CCGTGATCGGCCCAGGACCGCCAGA
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
There are no transcripts associated with this gene.

Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 542 Silent Mutation TCA,TCG S151S NP_001036097.1
NM_001042633.2 542 Silent Mutation TCA,TCG S157S NP_001036098.1
NM_033421.3 542 Missense Mutation ACC,GCC T154A NP_219489.1
NM_152897.2 542 Missense Mutation ACC,GCC T154A NP_690857.1
XM_005260608.4 542 Missense Mutation ACC,GCC T56A XP_005260665.1
XM_011529095.2 542 Missense Mutation ACC,GCC T6A XP_011527397.1
XM_011529096.2 542 Intron XP_011527398.1
XM_011529098.2 542 UTR 3 XP_011527400.1
XM_011529099.2 542 Intron XP_011527401.1
XM_017028120.1 542 Intron XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
There are no transcripts associated with this gene.

View Full Product Details