Product Details

SNP ID
rs114469224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38114076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTATAACACCGTGGGGGGAAAA[A/G]CTAACATGGATTTTTCTCTTGATTC
Phenotype
MIM: 604782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASH2L PubMed Links

Gene Details

Gene
ASH2L
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105214.2 Intron NP_001098684.1
NM_001261832.1 Intron NP_001248761.1
NM_001282272.1 Intron NP_001269201.1
NM_004674.4 Intron NP_004665.2
XM_005273682.1 Intron XP_005273739.1
XM_005273683.1 Intron XP_005273740.1
XM_006716412.1 Intron XP_006716475.1
XM_006716413.2 Intron XP_006716476.1

View Full Product Details