Product Details
- SNP ID
-
rs17208734
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:32393647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGTGAACCTCAGCTCTCAGCCT[C/T]GAAAACAAGGATGGCTGTACTACTC
- Phenotype
-
MIM: 606000
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTNL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117026188] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTNL2
- Gene Name
- butyrophilin like 2
- Gene
- HCG23
- Gene Name
- HLA complex group 23 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- LOC101929163
- Gene Name
- uncharacterized LOC101929163
There are no transcripts associated with this gene.
View Full Product Details