Product Details

SNP ID
hCV25933725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11353934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATTGTCTAGCTTCACAGGGAGG[A/C]TATGTTCAGAGAGCCTATTTGGGAG
Phenotype
MIM: 613771
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080503.2 Intron NP_001073972.2
Gene
PLPPR2
Gene Name
phospholipid phosphatase related 2
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
There are no transcripts associated with this gene.

View Full Product Details