Product Details

SNP ID
rs61744311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124326291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGAGACAGAGGGTGGCCGGTC[A/C]CCAGATGCCAGGCCCGGGGCCGGGG
Phenotype
MIM: 600848
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCOR2 PubMed Links

Gene Details

Gene
NCOR2
Gene Name
nuclear receptor corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077261.3 7416 Silent Mutation GGG,GGT G2365G NP_001070729.2
NM_001206654.1 7416 Silent Mutation GGG,GGT G2411G NP_001193583.1
NM_006312.5 7416 Silent Mutation GGG,GGT G2421G NP_006303.4

View Full Product Details