Product Details

SNP ID
rs1046228921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152994324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTAATACACAGACAATGCAACA[C/T]TGTATCATTTACAAATCCTTAATTT
Phenotype
MIM: 613542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTRF1L PubMed Links

Gene Details

Gene
MTRF1L
Gene Name
mitochondrial translational release factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114184.2 Intron NP_001107656.1
NM_001301047.2 Intron NP_001287976.1
NM_001301870.1 Intron NP_001288799.1
NM_001301871.1 Intron NP_001288800.1
NM_001301872.1 Intron NP_001288801.1
NM_019041.6 Intron NP_061914.3

View Full Product Details