Product Details

SNP ID
rs148875460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39529637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCGCAAAATCTGGCGAGGGTGC[C/G]GGACCGAGGGCCCATCATTTCAAGA
Phenotype
MIM: 604064 MIM: 610225
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATF4 PubMed Links

Gene Details

Gene
ATF4
Gene Name
activating transcription factor 4
There are no transcripts associated with this gene.

Gene
RPS19BP1
Gene Name
ribosomal protein S19 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194326.3 Intron NP_919307.1

View Full Product Details