Product Details

SNP ID
rs11111854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103951475 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGTAGTTTGATGCAATAAATTA[A/G]GAAATACATATTTAAGACTTATATT
Phenotype
MIM: 191175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135570.1 484 UTR 3 NP_001129042.1
XM_011538718.2 484 UTR 3 XP_011537020.1
XM_017019916.1 484 UTR 3 XP_016875405.1
XM_017019917.1 484 UTR 3 XP_016875406.1
Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.

View Full Product Details