Product Details

SNP ID
rs147268000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76567739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTAGGTAAGGTTGGGGGAGGGG[G/T]TGGTGAGAAAGGAACCACAGGTTCC
Phenotype
MIM: 610137
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNHG16 PubMed Links

Gene Details

Gene
SNHG16
Gene Name
small nucleolar RNA host gene 16
There are no transcripts associated with this gene.

Gene
SNORD1A
Gene Name
small nucleolar RNA, C/D box 1A
There are no transcripts associated with this gene.

Gene
SNORD1B
Gene Name
small nucleolar RNA, C/D box 1B
There are no transcripts associated with this gene.

Gene
SNORD1C
Gene Name
small nucleolar RNA, C/D box 1C
There are no transcripts associated with this gene.

Gene
ST6GALNAC2
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006456.2 Intron NP_006447.2
XM_005256954.3 Intron XP_005257011.1

View Full Product Details