Product Details

SNP ID
rs10838531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45924286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCCGACCAGCTCAAGGCAGGGGC[C/T]CAGCCCTTCCCCCCTCCCCTCAGCT
Phenotype
MIM: 609709 MIM: 603360 MIM: 608325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARGE2 PubMed Links

Gene Details

Gene
LARGE2
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300721.1 Intron NP_001287650.1
NM_001300722.1 Intron NP_001287651.1
NM_152312.4 Intron NP_689525.3
XM_005252787.2 Intron XP_005252844.1
XM_006718140.1 Intron XP_006718203.1
XM_006718141.3 Intron XP_006718204.1
XM_011519886.1 Intron XP_011518188.1
XM_011519888.1 Intron XP_011518190.1
XM_011519889.1 Intron XP_011518191.1
XM_011519890.1 Intron XP_011518192.1
XM_011519891.1 Intron XP_011518193.1
XM_011519892.2 Intron XP_011518194.1
XM_011519893.1 Intron XP_011518195.1
XM_017017173.1 Intron XP_016872662.1
Gene
PEX16
Gene Name
peroxisomal biogenesis factor 16
There are no transcripts associated with this gene.

Gene
PHF21A
Gene Name
PHD finger protein 21A
There are no transcripts associated with this gene.

View Full Product Details