Product Details

SNP ID
rs17452383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13388824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGCATGCTATGGTTTCAAGGAAG[A/G]TCAGCATCACAGCATTGGATTCTGT
Phenotype
MIM: 602550 MIM: 615933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARNTL PubMed Links

Gene Details

Gene
ARNTL
Gene Name
aryl hydrocarbon receptor nuclear translocator like
There are no transcripts associated with this gene.

Gene
BTBD10
Gene Name
BTB domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297741.1 2814 Intron NP_001284670.1
NM_001297742.1 2814 UTR 3 NP_001284671.1
NM_032320.6 2814 Intron NP_115696.2
XM_017018405.1 2814 UTR 3 XP_016873894.1
XM_017018406.1 2814 Intron XP_016873895.1
XM_017018407.1 2814 UTR 3 XP_016873896.1
XM_017018408.1 2814 UTR 3 XP_016873897.1
XM_017018409.1 2814 Intron XP_016873898.1
XM_017018410.1 2814 Intron XP_016873899.1
XM_017018411.1 2814 Intron XP_016873900.1

View Full Product Details