Product Details

SNP ID
rs34294471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22872742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCTCGTTCCTCCTCTAGGAGG[C/T]GCTCTGGCCCATCCAGACCGGATTA
Phenotype
MIM: 609921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267510.1 730 Silent Mutation GGC,GGT G13G XP_005267567.1

View Full Product Details