Product Details

SNP ID
rs144126376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111094959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTTGCAGTAAGTAAGTATGGA[C/G]TTTATTCTGAATTTATGCACATCTA
Phenotype
MIM: 609669
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WDR36 PubMed Links

Gene Details

Gene
WDR36
Gene Name
WD repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139281.2 Intron NP_644810.1

View Full Product Details