Product Details

SNP ID
rs116165962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74767425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAGTTAGTGGGACCTGAGGAC[C/T]CTGGGGCGCTGGGGTAGGGGGTGAG
Phenotype
MIM: 606544 MIM: 612497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFM2 PubMed Links

Gene Details

Gene
GFM2
Gene Name
G elongation factor mitochondrial 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281302.1 Intron NP_001268231.1
NM_032380.4 Intron NP_115756.2
NM_170681.2 Intron NP_733781.1
NM_170691.2 Intron NP_733792.1
XM_011543691.2 Intron XP_011541993.1
XM_017009986.1 Intron XP_016865475.1
Gene
NSA2
Gene Name
NSA2, ribosome biogenesis homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271665.1 Intron NP_001258594.1
NM_014886.4 Intron NP_055701.1
XM_011543098.1 Intron XP_011541400.1
XM_017008952.1 Intron XP_016864441.1
XM_017008953.1 Intron XP_016864442.1

View Full Product Details