Product Details

SNP ID
rs61740094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:234496179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGCTTCTCAATCTCTGCCAC[C/T]GGCAGCGACTTGGGCAGGTCCTGCT
Phenotype
MIM: 604787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4C PubMed Links

Gene Details

Gene
ARL4C
Gene Name
ADP ribosylation factor like GTPase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282431.1 875 Silent Mutation CCA,CCG P136P NP_001269360.1
NM_005737.3 875 Silent Mutation CCA,CCG P136P NP_005728.2

View Full Product Details