Product Details

SNP ID
hCV25939084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178239095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGCCCTCCCATTCCCCCACCCT[C/T]CCCTGCCTCTCCCAAGCCTGCCCTG
Phenotype
MIM: 610043 MIM: 614683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL23A1 PubMed Links

Gene Details

Gene
COL23A1
Gene Name
collagen type XXIII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173465.3 Intron NP_775736.2
Gene
PHYKPL
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
There are no transcripts associated with this gene.

View Full Product Details