Product Details
- SNP ID
-
rs34451260
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53307533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGGGGCAGAACAGGTGGTGGCC[C/T]TGGGAGAGGGTCCCAAGGGGCAGAG
- Phenotype
-
MIM: 605378
MIM: 611366
MIM: 604899
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AAAS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112987708] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AAAS
- Gene Name
- aladin WD repeat nucleoporin
- Gene
- C12orf10
- Gene Name
- chromosome 12 open reading frame 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021640.3 |
1734 |
Intron |
|
|
NP_067653.3 |
- Gene
- PFDN5
- Gene Name
- prefoldin subunit 5
There are no transcripts associated with this gene.
View Full Product Details