Product Details

SNP ID
rs61739885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134291597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGGTTTACTCTGGGTGGCACA[A/G]ACCAATTCAGCTTTCATGGTCTTCC
Phenotype
MIM: 610923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35B4 PubMed Links

Gene Details

Gene
SLC35B4
Gene Name
solute carrier family 35 member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032826.4 4556 UTR 3 NP_116215.1
XM_011516645.2 4556 Intron XP_011514947.1

View Full Product Details