Product Details

SNP ID
rs61743973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24558332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCTCTACTTATGGCTTTGTCCC[G/T]GGACACATCGCAGTGACACAACAGA
Phenotype
MIM: 300948 MIM: 300906
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCYT1B PubMed Links

Gene Details

Gene
PCYT1B
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163264.1 5129 UTR 3 NP_001156736.1
NM_001163265.1 5129 UTR 3 NP_001156737.1
NM_004845.4 5129 UTR 3 NP_004836.2
XM_017029977.1 5129 UTR 3 XP_016885466.1
Gene
PDK3
Gene Name
pyruvate dehydrogenase kinase 3
There are no transcripts associated with this gene.

View Full Product Details