Product Details

SNP ID
rs34489008
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:168183911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGTTACGTATAGATTAGCCGA[C/T]GAATTACATATGCCATCTCTCCCTG
Phenotype
MIM: 611807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIPRL PubMed Links

Gene Details

Gene
TIPRL
Gene Name
TOR signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031800.2 347 Silent Mutation GAC,GAT D38D NP_001026970.1
NM_152902.4 347 Silent Mutation GAC,GAT D38D NP_690866.1

View Full Product Details