Product Details

SNP ID
rs79179803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:567469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGACTTTGCCATTCCCACACAC[A/C]GTGGGGCCTTATTCTCTCTCTTGCA
Phenotype
MIM: 605754
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987233 PubMed Links

Gene Details

Gene
LOC107987233
Gene Name
uncharacterized LOC107987233
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017023921.1 Intron XP_016879410.1
Gene
NHLRC4
Gene Name
NHL repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301159.1 Intron NP_001288088.1
NM_176677.2 Intron NP_788850.1
Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.

Gene
PRR35
Gene Name
proline rich 35
There are no transcripts associated with this gene.

View Full Product Details