Product Details

SNP ID
rs11672479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39200599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAAAGGGCTCCTCCCTAACCCC[A/G]GGTGCTGTCACCACAGGTGTCCCAC
Phenotype
MIM: 615901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCCRP1 PubMed Links

Gene Details

Gene
NCCRP1
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001414.1 Intron NP_001001414.1
XM_011526906.2 Intron XP_011525208.1
Gene
SYCN
Gene Name
syncollin
There are no transcripts associated with this gene.

View Full Product Details