Product Details

SNP ID
rs116729446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29268132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTATTTTGGCCCGAGTGTCAC[A/G]TCGGGCGCTCTTTAGAGAGGACTGG
Phenotype
MIM: 608926 MIM: 133450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMID1 PubMed Links

Gene Details

Gene
EMID1
Gene Name
EMI domain containing 1
There are no transcripts associated with this gene.

Gene
EWSR1
Gene Name
EWS RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163285.1 65 UTR 5 NP_001156757.1
NM_001163286.1 65 UTR 5 NP_001156758.1
NM_001163287.1 65 UTR 5 NP_001156759.1
NM_005243.3 65 UTR 5 NP_005234.1
NM_013986.3 65 UTR 5 NP_053733.2
XM_005261389.3 65 Intron XP_005261446.1
XM_005261390.4 65 Intron XP_005261447.1
XM_011529995.2 65 Intron XP_011528297.1
XM_011529996.2 65 Intron XP_011528298.1
XM_011529997.1 65 Intron XP_011528299.1
XM_011529998.1 65 Intron XP_011528300.1
XM_011529999.2 65 Intron XP_011528301.1
XM_011530000.1 65 Intron XP_011528302.1
XM_011530001.2 65 Intron XP_011528303.1
XM_011530002.2 65 Intron XP_011528304.1
XM_017028644.1 65 Intron XP_016884133.1
XM_017028645.1 65 Intron XP_016884134.1
XM_017028646.1 65 Intron XP_016884135.1
XM_017028647.1 65 Intron XP_016884136.1
XM_017028648.1 65 Intron XP_016884137.1
XM_017028649.1 65 Intron XP_016884138.1
XM_017028650.1 65 Intron XP_016884139.1
XM_017028651.1 65 Intron XP_016884140.1
XM_017028652.1 65 Intron XP_016884141.1
XM_017028653.1 65 Intron XP_016884142.1
XM_017028654.1 65 Intron XP_016884143.1
XM_017028655.1 65 Intron XP_016884144.1
XM_017028656.1 65 Intron XP_016884145.1
XM_017028657.1 65 Intron XP_016884146.1
XM_017028658.1 65 Intron XP_016884147.1
XM_017028659.1 65 Intron XP_016884148.1
XM_017028660.1 65 Intron XP_016884149.1
XM_017028661.1 65 Intron XP_016884150.1
XM_017028662.1 65 Intron XP_016884151.1
XM_017028663.1 65 Intron XP_016884152.1
XM_017028664.1 65 Intron XP_016884153.1
XM_017028665.1 65 Intron XP_016884154.1
XM_017028666.1 65 Intron XP_016884155.1
Gene
RHBDD3
Gene Name
rhomboid domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006724224.3 65 Intron XP_006724287.1
XM_011530107.2 65 UTR 5 XP_011528409.1
XM_017028749.1 65 Intron XP_016884238.1
XM_017028750.1 65 Intron XP_016884239.1

View Full Product Details