Product Details
- SNP ID
-
rs35398622
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:35607782 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGTGTGCGTCCTCCTCCCCTCCG[T/C]CGTCTTCAGCGTCTATGTCCTGCAC
- Phenotype
-
MIM: 611922
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GJD4
PubMed Links
Gene Details
- Gene
- GJD4
- Gene Name
- gap junction protein delta 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153368.2 |
427 |
Missense Mutation |
GCC,GTC |
A90V |
NP_699199.2 |
View Full Product Details