Product Details

SNP ID
rs61741022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103494595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCTGGAGGCCGGAGATGCACCT[C/T]TGGCCGTGGAGTGGGCTGGGGGTGG
Phenotype
MIM: 603804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEURL1 PubMed Links

Gene Details

Gene
NEURL1
Gene Name
neuralized E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004210.4 Intron NP_004201.3
XM_005270269.3 Intron XP_005270326.1
XM_005270270.3 Intron XP_005270327.1
XM_011540331.2 Intron XP_011538633.1
XM_011540332.2 Intron XP_011538634.1
XM_011540333.2 Intron XP_011538635.1
XM_011540335.2 Intron XP_011538637.1
XM_017016909.1 Intron XP_016872398.1
Gene
NEURL1-AS1
Gene Name
NEURL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details