Product Details

SNP ID
rs73283603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56358454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACATCTCCAGCAGGTTGTAGACC[A/G]ACAGCCTTGGAGAAATACAGTATAG
Phenotype
MIM: 609177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 835 Silent Mutation GTC,GTT V219V NP_001005413.1
NM_007057.3 835 Silent Mutation GTC,GTT V266V NP_008988.2
NM_032997.2 835 Silent Mutation GTC,GTT V266V NP_127490.1
XM_017015605.1 835 Missense Mutation TCG,TTG S254L XP_016871094.1

View Full Product Details