Product Details
- SNP ID
-
rs7806638
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149839905 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCAAAATGTAGGGGGAAGAAGGA[C/T]TATAAAATATTGGGATTGATATTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSPO
PubMed Links
Gene Details
- Gene
- SSPO
- Gene Name
- SCO-spondin
There are no transcripts associated with this gene.
- Gene
- ZNF862
- Gene Name
- zinc finger protein 862
View Full Product Details