Product Details

SNP ID
rs55984737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17460537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCAGGAAGCCCTCCCTGCCCC[G/T]CCTCACCTCCTCAGATCATCCTCAA
Phenotype
MIM: 608791 MIM: 600691
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NXNL1 PubMed Links

Gene Details

Gene
NXNL1
Gene Name
nucleoredoxin-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138454.1 Intron NP_612463.1
Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
There are no transcripts associated with this gene.

View Full Product Details