Product Details

SNP ID
rs539698786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42619678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACGGGCAGGCCAGGCTGAACC[C/T]GGGGCCCCAGTGTGCGATGTGGGGA
Phenotype
MIM: 613213 MIM: 611520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5R3 PubMed Links

Gene Details

Gene
CYB5R3
Gene Name
cytochrome b5 reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000398.6 1225 UTR 3 NP_000389.1
NM_001129819.2 1225 UTR 3 NP_001123291.1
NM_001171660.1 1225 UTR 3 NP_001165131.1
NM_001171661.1 1225 UTR 3 NP_001165132.1
NM_007326.4 1225 UTR 3 NP_015565.1
Gene
POLDIP3
Gene Name
DNA polymerase delta interacting protein 3
There are no transcripts associated with this gene.

Gene
RNU12
Gene Name
RNA, U12 small nuclear
There are no transcripts associated with this gene.

View Full Product Details