Product Details

SNP ID
rs28647582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41747248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCCATGGGGCCCTAGGTCCTTC[C/T]CACTCGAGGCTCCAGGACTTCGAAT
Phenotype
MIM: 603851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHOX2B PubMed Links

Gene Details

Gene
PHOX2B
Gene Name
paired like homeobox 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003924.3 Intron NP_003915.2

View Full Product Details