Product Details

SNP ID
rs73646777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:71683794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATACAAAGGTAGAGGCCATCAGC[C/T]TTTGCCCCTAGAAGAGGAAAGTGAA
Phenotype
MIM: 605835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM2 PubMed Links

Gene Details

Gene
TMEM2
Gene Name
transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135820.1 5906 UTR 3 NP_001129292.1
NM_013390.2 5906 UTR 3 NP_037522.1
XM_005251869.4 5906 UTR 3 XP_005251926.1

View Full Product Details