Product Details

SNP ID
rs11895057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237085776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAGGTCCTCCAGGGCACCGAGA[A/G]AGCCGGCCAGAACGGCGGCGCCCTA
Phenotype
MIM: 616011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPS8 PubMed Links

Gene Details

Gene
COPS8
Gene Name
COP9 signalosome subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006710.4 336 UTR 5 NP_006701.1
NM_198189.2 336 UTR 5 NP_937832.1

View Full Product Details