Product Details

SNP ID
rs7654853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186534107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGTCTGACCTGGAGAACCAGG[A/G]TCCATATTCTCAGGTAACAGAAGAT
Phenotype
MIM: 600665
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTNR1A PubMed Links

Gene Details

Gene
MTNR1A
Gene Name
melatonin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005958.4 837 Missense Mutation ACC,ATC T212I NP_005949.1
XM_011532002.2 837 Missense Mutation ACC,ATC T127I XP_011530304.1

View Full Product Details