Product Details

SNP ID
rs61743131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104587852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCTCGAGGCGCGTGTGGTTCC[A/G]AGAACCCCCAGCGGTGACCGTCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf180 PubMed Links

Gene Details

Gene
C14orf180
Gene Name
chromosome 14 open reading frame 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008404.2 551 Missense Mutation CAA,CGA Q72R NP_001008404.1
NM_001286399.1 551 Missense Mutation CAA,CGA Q72R NP_001273328.1
NM_001286400.1 551 Missense Mutation CAA,CGA Q72R NP_001273329.1
XM_005267638.3 551 Missense Mutation CAA,CGA Q72R XP_005267695.1
XM_011536764.2 551 Missense Mutation CAA,CGA Q72R XP_011535066.1
Gene
TMEM179
Gene Name
transmembrane protein 179
There are no transcripts associated with this gene.

View Full Product Details