Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161707.1 | Intron | NP_001155179.1 | |||
NM_001301097.1 | Intron | NP_001288026.1 | |||
NM_032531.3 | Intron | NP_115920.1 | |||
XM_011543026.2 | Intron | XP_011541328.1 | |||
XM_011543027.2 | Intron | XP_011541329.1 | |||
XM_011543028.2 | Intron | XP_011541330.1 | |||
XM_011543030.2 | Intron | XP_011541332.1 | |||
XM_011543031.2 | Intron | XP_011541333.1 | |||
XM_011543032.2 | Intron | XP_011541334.1 | |||
XM_011543033.1 | Intron | XP_011541335.1 | |||
XM_017018418.1 | Intron | XP_016873907.1 | |||
XM_017018419.1 | Intron | XP_016873908.1 | |||
XM_017018420.1 | Intron | XP_016873909.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001254757.1 | Intron | NP_001241686.1 | |||
NM_001254758.1 | Intron | NP_001241687.1 | |||
NM_001254759.1 | Intron | NP_001241688.1 | |||
NM_006278.2 | Intron | NP_006269.1 | |||
XM_011542959.1 | Intron | XP_011541261.1 | |||
XM_011542960.2 | Intron | XP_011541262.1 | |||
XM_017018134.1 | Intron | XP_016873623.1 |