Product Details

SNP ID
rs34622520
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:32895665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCTGGGTGGCAAAGGTCATGG[C/T]GATGGAGGCCAAGAGGCCGAGGGCA
Phenotype
MIM: 142622
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
There are no transcripts associated with this gene.

Gene
TMEM54
Gene Name
transmembrane protein 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005270408.3 546 Missense Mutation ACC,GCC T117A XP_005270465.1

View Full Product Details