Product Details
- SNP ID
-
rs10876637
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54854838 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATTCAGTCAGATCTTACTTCAA[C/T]AATACCTTGGAACCCAAGTTTATTG
- Phenotype
-
MIM: 610857
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MUCL1
PubMed Links
Gene Details
- Gene
- MUCL1
- Gene Name
- mucin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_058173.2 |
|
Intron |
|
|
NP_477521.1 |
View Full Product Details