Product Details

SNP ID
rs200181105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6427291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGAGAAAGCTCACTAGGAGCC[C/T]GGGCTCCGGCCAGCCCTGCCACCCC
Phenotype
MIM: 603445 MIM: 608746 MIM: 610822
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KHSRP PubMed Links

Gene Details

Gene
KHSRP
Gene Name
KH-type splicing regulatory protein
There are no transcripts associated with this gene.

Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

Gene
SLC25A41
Gene Name
solute carrier family 25 member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321298.1 Intron NP_001308227.1
NM_173637.3 Intron NP_775908.2
XM_011527926.1 Intron XP_011526228.1

View Full Product Details