Product Details

SNP ID
rs35659581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4267945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTAAAGGTAGGGTTCTTGCTGA[G/T]TTTCTTGTTAAAGATGACCAGGAGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145725.1 1258 Missense Mutation ATC,CTC I362L NP_001139197.1
NM_017816.2 1258 Missense Mutation ATC,CTC I362L NP_060286.1
XM_011513505.1 1258 Missense Mutation ATC,CTC I362L XP_011511807.1
XM_011513506.2 1258 Missense Mutation ATC,CTC I362L XP_011511808.1

View Full Product Details