Product Details
- SNP ID
-
rs62622789
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137613028 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGTAAGTTTGGGAGCCAGTTCC[C/T]GAGTGGTGACCCCTGGGGGCAGCCC
- Phenotype
-
MIM: 607001
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARRDC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76318189] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARRDC1
- Gene Name
- arrestin domain containing 1
- Gene
- ARRDC1-AS1
- Gene Name
- ARRDC1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- EHMT1
- Gene Name
- euchromatic histone lysine methyltransferase 1
There are no transcripts associated with this gene.
View Full Product Details